missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RELT/TNFRSF19L Rabbit anti-Human, Clone: 135, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89556-100UL
This item is not returnable.
View return policy
Description
RELT/TNFRSF19L Monoclonal specifically detects RELT/TNFRSF19L in Human samples. It is validated for ELISA, Sandwich ELISA Detection.
Specifications
| RELT/TNFRSF19L | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 135 | |
| ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000 | |
| Receptor expressed in lymphoid tissues, RELT tumor necrosis factor receptor, TNFRSF19LFLJ14993, tumor necrosis factor receptor superfamily member 19L, tumor necrosis factor receptor superfamily, member 19-like | |
| Recombinant Human RELT/TNFRSF19L protein | |
| 100 μg | |
| Cancer | |
| 84957 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction