missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RELT/TNFRSF19L Antibody (135), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89556CL1
This item is not returnable.
View return policy
Description
RELT/TNFRSF19L Monoclonal antibody specifically detects RELT/TNFRSF19L in Human samples. It is validated for ELISA,Sandwich ELISA Detection
Specifications
| RELT/TNFRSF19L | |
| Monoclonal | |
| CoraFluor 1 | |
| Receptor expressed in lymphoid tissues, RELT tumor necrosis factor receptor, TNFRSF19LFLJ14993, tumor necrosis factor receptor superfamily member 19L, tumor necrosis factor receptor superfamily, member 19-like | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human RELT/TNFRSF19L (Accession#: NP_116260.2; Met 1-Ala 160). | |
| 0.1 mL | |
| Cancer | |
| 84957 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Sandwich ELISA Detection | |
| 135 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction