missing translation for 'onlineSavingsMsg'
Learn More

RELT/TNFRSF19L Antibody (111), DyLight 550 , Novus Biologicals™

Product Code. 18412156 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18412156 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18412156 Supplier Novus Biologicals Supplier No. NBP289555R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

RELT/TNFRSF19L Monoclonal antibody specifically detects RELT/TNFRSF19L in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen RELT/TNFRSF19L
Applications ELISA
Classification Monoclonal
Clone 111
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias Receptor expressed in lymphoid tissues, RELT tumor necrosis factor receptor, TNFRSF19LFLJ14993, tumor necrosis factor receptor superfamily member 19L, tumor necrosis factor receptor superfamily, member 19-like
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human RELT/TNFRSF19L (NP_116260.2; Met1-Ala160).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 84957
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.