missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RELM gamma Monoclonal Antibody (395310)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561204

Product Code. 30536006

  • £382.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

RELMgamma belongs to a unique family of tissue-specific cytokines termed FIZZ (found in inflammatory zone) and RELM. The other three known members of this family, resistin, RELMalpha, and RELMbeta, are 85-94 amino acid, secreted proteins sharing a conserved C-terminal domain, characterized by 10 cysteine residues with a unique spacing motif of C-X11-C-X8-C-X-C-X3-C-X10-C-X-C-X-C-X9-C-C. RELMgamma is most closely related to RELMalpha, but is distinctly secreted in the bone marrow, spleen, lung, and peripheral blood granulocytes. The physiological role of RELMgamma may include the promotion or regulation of promyelocytic differentiation, although the specific molecular targets of RELMgamma have not been identified.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RELM gamma
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
G3V686
Mouse
Protein A/G
RUO
288135
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Western Blot
395310
Unconjugated
Retnlg
Retnlg
Mouse myeloma cell line NS0-derived recombinant rat RELM gamma , Asp24-Ser111.
100 μg
Primary
Rat
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.