missing translation for 'onlineSavingsMsg'
Learn More

RELM beta, Rat anti-Mouse, Clone: MRB 46L, Novus Biologicals™

Rat Monoclonal Antibody

Brand:  Novus Biologicals NBP2-80033-100UG

 View more versions of this product

Product Code. 18253638

  • £376.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

RELM beta Monoclonal specifically detects RELM beta in Mouse samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RELM beta
Monoclonal
1 mg/mL
Western Blot, ELISA
C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 2, CCRG, Colon and small intestine-specific cysteine-rich protein, Colon carcinoma-related gene protein, cysteine-rich secreted A12-alpha-like protein 1, Cysteine-rich secreted protein A12-alpha-like 1, Cysteine-rich secreted protein FIZZ2, FIZZ1, FIZZ2RELM-beta, found in inflammatory zone 1, HXCP2XCP2, RELMb, RELMbeta, resistin like beta, resistin-like beta, RETNL2
Rat
Protein G purified
RUO
Primary
Recognizes mouse RELM beta. Detects a band of ∽10kDa by Western blot. Does not cross-react with human RELM beta.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2a κ
Western Blot, ELISA
MRB 46L
Unconjugated
0.2 um-filtered solution in PBS, pH 7.4
RETNLB
Recombinant mouse RELM beta.
100 μg
Cancer
84666
Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.