missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RELM beta Antibody (MRB 46L), DyLight 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-80033G
This item is not returnable.
View return policy
Description
RELM beta Monoclonal antibody specifically detects RELM beta in Mouse samples. It is validated for Western Blot, ELISA
Specifications
| RELM beta | |
| Monoclonal | |
| DyLight 488 | |
| C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 2, CCRG, Colon and small intestine-specific cysteine-rich protein, Colon carcinoma-related gene protein, cysteine-rich secreted A12-alpha-like protein 1, Cysteine-rich secreted protein A12-alpha-like 1, Cysteine-rich secreted protein FIZZ2, FIZZ1, FIZZ2RELM-beta, found in inflammatory zone 1, HXCP2XCP2, RELMb, RELMbeta, resistin like beta, resistin-like beta, RETNL2 | |
| Recombinant mouseRELM beta. | |
| 0.1 mL | |
| Cancer | |
| 84666 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, ELISA | |
| MRB 46L | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction