missing translation for 'onlineSavingsMsg'
Learn More

RELL2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13278458
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13278458 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13278458 Supplier Invitrogen Supplier No. PA523910

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

RELL2 Polyclonal Antibody

RELL2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen RELL2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RELL2
Gene Accession No. Q8BRJ3, Q8NC24
Gene Alias 4631403P03Rik; C5orf16; ependolin; receptor expressed in lymphoid tissues like 2; Rell2; RELT like 2; RELT-like 2; RELT-like protein 2; UNQ9423/PRO34565
Gene Symbols RELL2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 132-160 amino acids from the central region of human RELL2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225392, 285613
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.