missing translation for 'onlineSavingsMsg'
Learn More

Relebactam, MedChemExpress

Product Code. 16487166
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487166 100 mg 100mg
16467178 10 mg 10mg
16413546 25 mg 25mg
16469707 5 mg 5mg
16403207 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16487166 Supplier Medchem Express Supplier No. HY16752

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Relebactam is a diazabicyclooctane inhibitor with activity against a wide spectrum of β-lactamases, including class A (extended-spectrum β-lactamases and KPC) and class C (AmpC) enzymes. Relebactam shows antibacterial activity.

Relebactam is a diazabicyclooctane inhibitor with activity against a wide spectrum of β-lactamases, including class A (extended-spectrum β-lactamases and KPC) and class C (AmpC) enzymes. Relebactam shows antibacterial activity.

Specifications

Chemical Name or Material Relebactam
Physical Form Solid
CAS 1174018-99-5
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C12H20N4O6S
For Use With (Application) COVID-19-immunoregulation
Synonym MK-7655
Solubility Information H2O : 50 mg/mL (143.52 mM; Need ultrasonic)
SMILES O=S(ON1[C@]2([H])CC[C@@H](C(NC3CCNCC3)=O)[N@@](C2)C1=O)(O)=O
Molecular Weight (g/mol) 348.38
Formula Weight 348.38
Percent Purity 99.56%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.