missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Relaxin R1/LGR7 Antibody (933344), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8898CL1
This item is not returnable.
View return policy
Description
Relaxin R1/LGR7 Monoclonal antibody specifically detects Relaxin R1/LGR7 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF
Specifications
| Relaxin R1/LGR7 | |
| Monoclonal | |
| CoraFluor 1 | |
| leucine-rich repeat-containing G protein-coupled receptor 7, Leucine-rich repeat-containing G-protein coupled receptor 7, LGR7.1, LGR7.10, LGR7LGR7.2, MGC138347, MGC142177, Relaxin family peptide receptor 1, relaxin receptor 1, relaxin/insulin-like family peptide receptor 1, RXFPR1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Relaxin R1, Met1-Ser398, Accession # Q9HBX9 | |
| 0.1 mL | |
| GPCR | |
| 59350 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF | |
| 933344 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction