missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Relaxin-3 Antibody (MM0526-7G14), mFluor Violet 610 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Relaxin-3 Monoclonal antibody specifically detects Relaxin-3 in Human samples. It is validated for Western Blot, Neutralization
Specifications
Specifications
| Antigen | Relaxin-3 |
| Classification | Monoclonal |
| Conjugate | mFluor Violet 610 SE |
| Formulation | 50mM Sodium Borate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?