missing translation for 'onlineSavingsMsg'
Learn More

Relaxin-3 Antibody (MM0526-7G14), Allophycocyanin, Novus Biologicals™

Product Code. 18400302 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400302 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400302 Supplier Novus Biologicals Supplier No. NBP211866APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Relaxin-3 Monoclonal antibody specifically detects Relaxin-3 in Human samples. It is validated for Western Blot, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen Relaxin-3
Applications Western Blot, Neutralization
Classification Monoclonal
Clone MM0526-7G14
Conjugate APC
Dilution Western Blot, Block/Neutralize
Formulation PBS
Gene Alias H3, insl7, relaxin 3, RXN3, ZINS4
Host Species Mouse
Immunogen Human recombinant Relaxin-3
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 117579
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.