missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Relaxin-3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41317-0.025mg
This item is not returnable.
View return policy
Description
Relaxin-3 Polyclonal specifically detects Relaxin-3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Relaxin-3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| RLN3 | |
| Antibody was raised against a 19 amino acid peptide near the center of human RLN3. The immunogen is located within amino acids 80 - 130 of RLN3. Amino Acid Squence: LALTKSPQAFYRGRPSWQG | |
| Affinity Purified | |
| RUO | |
| Primary | |
| RLN3 antibody is human specific. RLN3 antibody is predicted to not cross-react with other members of the Relaxin protein family. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| H3, insl7, relaxin 3, RXN3, ZINS4 | |
| Rabbit | |
| 16 kDa | |
| 0.025 mg | |
| Neuroscience, Neurotransmission | |
| 117579 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction