missing translation for 'onlineSavingsMsg'
Learn More

Relaxin-1 Antibody (361505) [FITC], Novus Biologicals™

Product Code. 30734346 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734346 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734346 Supplier Novus Biologicals Supplier No. FAB32571F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Relaxin-1 Monoclonal antibody specifically detects Relaxin-1 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Relaxin-1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 361505
Conjugate FITC
Formulation PBS
Gene Alias bA12D24.3.1, bA12D24.3.2, H1, preprorelaxin H1, prorelaxin H1, relaxin 1, relaxin 1 (H1), relaxin H1, RLXH1
Host Species Rat
Immunogen E. coli-derived recombinant human Relaxin-1, Lys26-Cys185, Accession # P04808
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6013
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.