missing translation for 'onlineSavingsMsg'
Learn More

Relative Density Standard 15°C, UKAS ISO 17025 / ISO 17034, 500 mL, nominal Relative Density 15/4 0.8494 at 15 °C, Paragon Scientific™

Product Code. 16020310
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 16020310

Brand: Paragon Scientific RDEN1505

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Paragon ISO 17025 / ISO 17034 Relative Density standards are designed for the calibration or verification of instruments used to measure density and relative density.ISO 17025 / ISO 17034 certification under UKAS ensures full traceability to national standards.

  • Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Certified in accordance with primary level ASTM D1480 methodology
  • The Relative density was calculated by dividing the density, as obtained from ASTM D1480, by the density of water at the reference temperature.
  • The density of water was taken from the table denoted 'Density of Water' found in ASTM D1480 and confirms to the International Temperature Scale (ITS 90).
  • Supplied in tamper evident packaging with a 2 year shelf life
  • Manufactured in the United Kingdom

Specifications

Quantity 500 mL
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Kerosine : distillates, hydrotreated light, White mineral oil, petroleum

Nominal values quoted. Certified Values will vary from batch to batch.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.