missing translation for 'onlineSavingsMsg'
Learn More

RELA, Rabbit anti-Human, Clone: C8, Abnova™

Rabbit Monoclonal Antibody

Brand:  Abnova MAB18975.200uL

Product Code. 16160350

  • £752.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

RELA
Monoclonal
Unconjugated
In PBS, pH 7.4 (50% glycerol, 0.02% sodium azide, 0.1% BSA).
MGC131774/NFKB3/p65
Rabbit
200 μL
Primary
Human
Antibody
IgG1 κ
Western Blot
C8
Western Blot (1 ug/mL - 0.001 ug/mL) The optimal working dilution should be determined by the end user.
v-rel reticuloendotheliosis viral oncogene homolog A (avian)
RELA
A synthetic phosphopeptide corresponding to residues surrounding S536 of human RELA.
RUO
5970
Store at -20°C.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.