missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ RelA/NFkB p65 [p Ser276] Inhibitor Peptide Set

Product Code. 18122254 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18122254 5 mg 5mg
18711134 - -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18122254 Supplier Novus Biologicals™ Supplier No. NBP2265055MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Applications: In vivo assay, Functional (Inhibition), Block/Neutralize

TRUSTED_SUSTAINABILITY

Specifications

Host Species Human, Mouse, Rat, Bovine, Canine, Mammal
Components NF-kB p65 (Ser276) Inhibitor peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKKQLRRPSDRELSE (p65 sequence: QLRRPSDRELSE). Molecular weight: 3830; Antennapedia Control peptide: 2 x 1mg (lyophilized) DRQIKIWFQNRRMKWKK. Molecular weight: 2361
For Use With (Application) Blocking/Neutralizing, Functional (Inhibition), In vivo assay
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 5 mg
Product Type RelA/NFkB p65 [p Ser276] Inhibitor Peptide Set
Inhibitors RelA/NFkB p65 [p Ser276]
Form Lyophilized

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.