missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RelA/NFkB p65 NLS Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-24541
This item is not returnable.
View return policy
Description
RelA/NFkB p65 NLS Polyclonal specifically detects RelA/NFkB p65 NLS in Human, Mouse, Bovine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| RelA/NFkB p65 NLS | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| RELA | |
| A portion of the NFkB p65 NLS nuclear localization signal (NLS) (amino acids DTDDRHRIEEKRKRKT) was used as the immunogen for this antibody. | |
| 0.1 mg | |
| Primary | |
| The amino acid sequence used as immunogen for this phospho antibody is 100% homologous in human, mouse, rat, bull, chicken, chimp, Drosophila, dog, rhesus monkey, Xenopus and zebrafish. It is also 100% homologous with MEK2, with the phospho-serines at positions 222 and 226. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1-3 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
| MGC131774, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65 NLS, p65 Nuclear Localization Sequence, p65-NLS, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 5970 | |
| Human, Mouse, Bovine, Equine, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction