missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RelA/NFkB p65 NLS Antibody, Allophycocyanin, Novus Biologicals™
Description
RelA/NFkB p65 NLS Polyclonal antibody specifically detects RelA/NFkB p65 NLS in Human, Mouse, Bovine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | RelA/NFkB p65 NLS |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | APC |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | PBS |
| Gene Alias | MGC131774, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65 NLS, p65 Nuclear Localization Sequence, p65-NLS, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65 |
| Host Species | Rabbit |
| Immunogen | A portion of the NFkB p65 NLS nuclear localization signal (NLS) (amino acids DTDDRHRIEEKRKRKT) was used as the immunogen for this antibody. |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?