missing translation for 'onlineSavingsMsg'
Learn More

RelA/NFkB p65 Antibody (532301), PE, Novus Biologicals™

Product Code. 30026432 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30026432 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30026432 Supplier Novus Biologicals Supplier No. FAB5078P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RelA/NFkB p65 Monoclonal antibody specifically detects RelA/NFkB p65 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen RelA/NFkB p65
Applications Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF, KnockDown
Classification Monoclonal
Clone 532301
Conjugate PE
Dilution Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready, Knockout Validated
Formulation PBS
Gene Alias MGC131774, nf kb p65, NFkB p65, NF-kB p65, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65
Host Species Mouse
Immunogen E. coli-derived recombinant human RelA/NF kappa B p65 isoform 1, Asn456-Ser551, Accession # Q04206
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5970
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.