missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RelA/NFkB p65 Antibody (532301) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5078JF669
This item is not returnable.
View return policy
Description
RelA/NFkB p65 Monoclonal antibody specifically detects RelA/NFkB p65 in Human,Mouse,Rat samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),Immunocytochemistry,CyTOF,Gene Knock-Out
Specifications
| RelA/NFkB p65 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| MGC131774, nf kb p65, NFkB p65, NF-kB p65, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65 | |
| E. coli-derived recombinant human RelA/NF kappa B p65 isoform 1, Asn456-Ser551, Accession # Q04206 | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF, Gene Knock-Out | |
| 532301 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 5970 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction