missing translation for 'onlineSavingsMsg'
Learn More

rel-trans-6-(Dimethylamino)-3-(4-hydroxycyclohexyl)-1-methyl-1,3,5-Triazine-2,4(1H,3H)-dione, TRC

Product Code. 30382672
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382672 10 mg -
30370511 50 mg 50mg
30380053 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382672 Supplier Toronto Research Chemicals Supplier No. TRCD48558010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72576-13-7
Molecular Formula C12 H20 N4 O3
Molecular Weight (g/mol) 268.3122
Synonym 1,3,5-Triazine-2,4(1H,3H)-dione, 6-(dimethylamino)-3-(4-hydroxycyclohexyl)-1-methyl-, 6-(Dimethylamino)-3-(4-hydroxycyclohexyl)-1-methyl-1,3,5-triazine-2,4(1H,3H)-dione
IUPAC Name 6-(dimethylamino)-3-(4-hydroxycyclohexyl)-1-methyl-1,3,5-triazine-2,4-dione
SMILES CN(C)C1=NC(=O)N(C2CCC(O)CC2)C(=O)N1C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C12H20N4O3/c1-14(2)10-13-11(18)16(12(19)15(10)3)8-4-6-9(17)7-5-8/h8-9,17H,4-7H2,1-3H3
Formula Weight 268.1535
Chemical Name or Material Hexazinone metabolite A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.