missing translation for 'onlineSavingsMsg'
Learn More

rel-trans-1-(4-Hydroxycyclohexyl)-3-methyl-1,3,5-triazinane-2,4,6-trione, TRC

Product Code. 30383027
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383027 1 mg -
30385864 5 mg -
30375107 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383027 Supplier Toronto Research Chemicals Supplier No. TRCD4855851MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C10 H15 N3 O4
Molecular Weight (g/mol) 241.244
Synonym rel-trans-4-hydroxy-hexazinone, N, N-dimethyl hexazinone rel-trans-3-(4-Hydroxycyclohexyl)-1-methyl-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, rel-trans-1-(4-Hydroxycyclohexyl)-3-methyl-1,3,5-triazinane-2,4,6-trione, rel-trans-1-(4-Hydroxycyclohexyl)-3-methyl-1,3,5-triazine-2,4,6(1H,3H,5H)-trione
IUPAC Name 1-(4-hydroxycyclohexyl)-3-methyl-1,3,5-triazinane-2,4,6-trione
SMILES CN1C(=O)NC(=O)N([C@@H]2CC[C@@H](O)CC2)C1=O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C10H15N3O4/c1-12-8(15)11-9(16)13(10(12)17)6-2-4-7(14)5-3-6/h6-7,14H,2-5H2,1H3,(H,11,15,16)/t6-,7-
Formula Weight 241.106
Chemical Name or Material rel-trans-1-(4-Hydroxycyclohexyl)-3-methyl-1,3,5-triazinane-2,4,6-trione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.