missing translation for 'onlineSavingsMsg'
Learn More

rel-(3R,5R)-Fluvastatin Sodium, TRC

Product Code. 30401422
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401422 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401422 Supplier Toronto Research Chemicals Supplier No. TRCF6012155MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3RS,5RS,6E)-7-(3-(4-Fluorophenyl)-1-methylethyl-1H-indol-2-yl)-3,5-dihydroxyhept-6-enoic Acid Sodium Salt
CAS 93957-58-5
Molecular Formula C24 H25 F N O4 . Na
Quantity 5 mg
Synonym 6-Heptenoic acid, 7-[3-(4-fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxy-, sodium salt (1:1), (3R,5R,6E)-rel-, 6-Heptenoic acid, 7-[3-(4-fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxy-, monosodium salt, [R*,R*-(E)]- (9CI), 6-Heptenoic acid, 7-[3-(4-fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxy-, monosodium salt, [R*,R*-(E)]-(±)-, (3RS,5RS,6E)-7-[3-(4-Fluorophenyl)-1-methylethyl-1H-indol-2-yl]-3,5-dihydroxyhept-6-enoic acid sodium salt, Fluvastatin sodium anti-isomer, Fluvastatin EP Impurity A Sodium Salt
InChI Formula InChI=1S/C24H26FNO4.Na/c1-15(2)26-21-6-4-3-5-20(21)24(16-7-9-17(25)10-8-16)22(26)12-11-18(27)13-19(28)14-23(29)30;/h3-12,15,18-19,27-28H,13-14H2,1-2H3,(H,29,30);/q;+1/p-1/b12-11+;/t18-,19+;/m1./s1
SMILES [Na+].CC(C)n1c(\C=C\[C@@H](O)C[C@H](O)CC(=O)[O-])c(c2ccc(F)cc2)c3ccccc13
IUPAC Name sodium;(E,3S,5S)-7-[3-(4-fluorophenyl)-1-propan-2-ylindol-2-yl]-3,5-dihydroxyhept-6-enoate
Molecular Weight (g/mol) 433.45
Formula Weight 433.1665
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.