missing translation for 'onlineSavingsMsg'
Learn More

rel-(2R,3aR,5S,6S,6aS)-6-(Hydroxy-2,2-di(thiophen-2-yl)acetoxy)-4,4-dimethylhexahydro-2H-2,5-methanofuro[3,2-b]pyrrol-4-ium Bromide, TRC

Product Code. 30403174
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
50 mg
Unit Size:
2.5mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403174 50 mg 50mg
30388937 25 mg 25mg
30407359 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30403174 Supplier Toronto Research Chemicals Supplier No. TRCH80524050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rel-(2R,3aR,5S,6S,6aS)-6-(Hydroxy-2,2-di(thiophen-2-yl)acetoxy)-4,4-dimethylhexahydro-2H-2,5-methanofuro[3,2-b]pyrrol-4-ium Bromide
Analyte or Component Names rel-(2R,3aR,5S,6S,6aS)-6-(Hydroxy-2,2-di(thiophen-2-yl)acetoxy)-4,4-dimethylhexahydro-2H-2,5-methanofuro[3,2-b]pyrrol-4-ium Bromide
Shipping Condition Room Temperature
Molecular Formula C19 H21 N O4 S2 . Br
Synonym Tiotoropium Bromide EP Impurity N
InChI Formula InChI=1S/C19H21NO4S2.BrH/c1-20(2)12-9-11-10-13(20)17(16(12)23-11)24-18(21)19(22,14-5-3-7-25-14)15-6-4-8-26-15;/h3-8,10-13,16-17,22H,9H2,1-2H3;1H/q+1;/p-1/t11-,12-,13+,16+,17+;/m1./s1
Solution Type Neat
SMILES [Br-].C[N]1(C)[C@@H]2C[C@@H]3[CH+][C@H]1[C@H](OC(=O)C(O)(c4cccs4)c5cccs5)[C@H]2O3
Industry Type Pharmaceutical
Molecular Weight (g/mol) 471.408
Quantity 50 mg
Formula Weight 470.01
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.