missing translation for 'onlineSavingsMsg'
Learn More

rel-10-[(1R,2S)-2,3-Dihydroxy-1-(hydroxymethyl)propyl]-1,4,7,10-tetraazacyclododecane-1,4,7-triacetic Acid (>85%), TRC

Product Code. 30383199
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383199 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383199 Supplier Toronto Research Chemicals Supplier No. TRCD4795405MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rel-10-[(1R,2S)-2,3-Dihydroxy-1-(hydroxymethyl)propyl]-1,4,7,10-tetraazacyclododecane-1,4,7-triacetic Acid
CAS 151852-61-8
Molecular Formula C18 H34 N4 O9
Quantity 5 mg
Synonym 1,4,7,10-Tetraazacyclododecane-1,4,7-triacetic acid, 10-[(1R,2S)-2,3-dihydroxy-1-(hydroxymethyl)propyl]-, rel-, 1,4,7,10-Tetraazacyclododecane-1,4,7-triacetic acid, 10-[2,3-dihydroxy-1-(hydroxymethyl)propyl]-, (R*,S*)-(±)-, rel-10-[(1R,2S)-2,3-Dihydroxy-1-(hydroxymethyl)propyl]-1,4,7,10-tetraazacyclododecane-1,4,7-triacetic acid, 1,4,7,10-Tetraazacyclododecane-1,4,7-triacetic acid, 10-[2,3-dihydroxy-1-(hydroxymethyl)propyl]-, (R*,S*)-, 2,2',2Ê1'-[10-[(1RS,2SR)-2,3-Dihydroxy-1-(hydroxymethyl)propyl]-1,4,7,10-tetraazacyclododecane-1,4,7-triyl]triacetic acid
InChI Formula InChI=1S/C18H34N4O9/c23-12-14(15(25)13-24)22-7-5-20(10-17(28)29)3-1-19(9-16(26)27)2-4-21(6-8-22)11-18(30)31/h14-15,23-25H,1-13H2,(H,26,27)(H,28,29)(H,30,31)/t14-,15-/m1/s1
SMILES OC[C@@H](O)[C@@H](CO)N1CCN(CC(=O)O)CCN(CC(=O)O)CCN(CC(=O)O)CC1
IUPAC Name 2-[4,10-bis(carboxymethyl)-7-[(2R,3S)-1,3,4-trihydroxybutan-2-yl]-1,4,7,10-tetrazacyclododec-1-yl]acetic acid
Molecular Weight (g/mol) 450.48
Formula Weight 450.2326
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.