missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Fan, Reinforced

Product Code. 16404059 Shop All BINDER Products
Change view
Click to view available options
For Use With (Equipment):
BINDER Chambers FD 720
BINDER Chambers FED 720
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
16404059 BINDER Chambers FD 720 Each
16474439 BINDER Chambers FED 720 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16404059 Supplier BINDER™ Supplier No. 80121062
  • ONLINE EXCLUSIVE PRICE
    £2730.00
    £2676.00 / Pack of 1
    Save £54.00
    1% Off
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Reinforced fan for increased air change rates, including up-rated heat output for faster heating up time and improved recovery time up to 300°C; not in conjunction with load management option.

  • 400 V option model
  • Increases air change rates
  • Uprated heat output for faster heating up time and improved recovery time
  • For use with temperatures up to 300°C
  • Not for use in conjunction with load management option

Specifications

Description Fan, reinforced - 400 V option model, for increased air change rates, incl. uprated heat output for faster heating up time and improved recovery time; max. 300°C; not in conjunction with load management option
For Use With (Equipment) BINDER Chambers FD 720
Temperature (Metric) 300°C (Max.)
Type Fan
For Use With (Application) Increasing Air Change Rates, Including Uprated Heat Output 2200 W
Voltage 400 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.