Learn More
Thermo Scientific™ Reinforced Clostridial Medium (RCM) (Dehydrated)
Description
Cultivate and enumerate clostridia and other anaerobes occurring in food and pathological samples with Thermo Scientific™ Oxoid™ Reinforced Clostridial Medium. A semi-solid, highly nutritious medium can be used as an enrichment medium to optimize the growth of clostridia before sub-culturing to solid agar.
Use Reinforced Clostridial Medium (RCM) for the isolation and cultivation of anaerobic organisms occurring in a variety of habitats, including food and pathological specimens.
- Superior fertility: the medium produced the highest viable count when compared to other media tested by Hirsch and Grinstead 1
- Differential: can be made differential for sulphite-reducing clostridia by the addition of sodium sulphite and ferric citrate2
Reinforced Clostridial Medium (RCM) was designed by Hirsch and Grinstead1 for the cultivation and enumeration of clostridia. They showed that the medium was more fertile and enabled growth to be initiated from small inocula more readily than five other media tested. In a further comparison, the highest viable count obtainable was the criterion used, and again, RCM proved superior. Compared with the spleen infusion medium of Mundt and Jones3, RCM gave somewhat higher counts (Gibbs and Hirsch4).
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Content And Storage | 10°C to 30°C |
| Form | Powder |
| Media Type | Reinforced Clostridial Medium |
| Product Type | Dehydrated Culture Media |
| Quantity | 5 kg |
For Laboratory Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.