missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reinecke Salt, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly bottle
Unit Size:
100g
25g
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
Specifications
| Melting Point | 268°C to 272°C |
| Physical Form | Solid |
| Quantity | 25 g |
| Linear Formula | NH4[Cr(NH3)2(SCN)4] |
| UN Number | NONH for all modes of transport |
| Formula Weight | 336.43g/mol |
| Packaging | Poly bottle |
| Chemical Name or Material | Reinecke salt |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction