missing translation for 'onlineSavingsMsg'
Learn More

Reinecke Salt, ACS Reagent, ≥93.0%, Solstice

Product Code. 15661650 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15661650 25 g Glass Bottle 25g
15651650 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15661650 Supplier Honeywell-Fluka Supplier No. 18368725G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS reagent,≥93.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13573-16-5
MDL Number MFCD00066638

Specifications

Melting Point 272°C
Physical Form Solid
Quantity 25 g
Linear Formula NH4[Cr(NH3)2(SCN)4]
UN Number NONH for all modes of transport
Formula Weight 336.43g/mol
Percent Purity ≥93.0%
Grade ACS Reagent
Packaging Glass Bottle
Chemical Name or Material Reinecke Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.