missing translation for 'onlineSavingsMsg'
Learn More

Reichert™ Goldberg Clinical Handheld TS Meter Refractometer

Product Code. 11998224
Click to view available options
:
For Total Solids
This item is not returnable. View return policy

Product Code. 11998224

Brand: Reichert™ 1310400A

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Offer reading accuracy better than most benchtop laboratory refractometers

  • Provides rapid, reliable measurements
  • For measuring urione specific gravity, serum, plasma, protein, refractive index
  • Incorporates the Goldberg method, recognized as the most accurate and precise for handheld refractometer
  • Ideal for use in quality control, research, and clinical laboratories
  • Features a hermetically sealed liquid-filled optical prism that changes index with temperature to compensate instantly and automatically for temperature changes
  • Heavy-duty anodized aluminum body
  • Magnified and focusable scales feature easy-to-read markings and divisions

Specifications

Type Handheld Clinical TS Meter Refractometer
Scale Range 1 to 1.035 Urine Specific Gravity, 2.5 to 15 g/100 mL Serum Protein
Refractive Index 0 to 320
Accuracy ±0.001 USG, ±0.1g/mL Serum Protein, ±2nD
Resolution 0.0001
Increments 0.0001 sp. gr., 0.1g/100mL, 2 (n-nD x 104)
Dimensions (L x W x H) 1 x 1 x 6.2 in. (25.4 x 25.4 x 157.4 mm)
Height (English) 6.2 in.
Height (Metric) 157.4 mm
Length (English) 1 in.
Length (Metric) 25.4 mm
Width (English) 1 in.
Width (Metric) 25.4 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.