missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Oxoid Rehydration Fluid (10 x 1.2ml vials)
Brand: Thermo Scientific Oxoid R4710000
49.43 GBP valid until 2025-12-31
BEST PRICE on Promo! Use promo code "24137" to get your promotional price.
Rehydration Fluid (10 x 1.2ml vials)
This item is not returnable.
View return policy
Description
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction