missing translation for 'onlineSavingsMsg'
Learn More

Rehmannioside D, MedChemExpress

Product Code. 16469188
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16469188 10 mg 10mg
16461598 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16469188 Supplier Medchem Express Supplier No. HYN0912
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Rehmannioside D is a carotenoid glycoside.

Rehmannioside D is a carotenoid glycoside.

Specifications

Chemical Name or Material Rehmannioside D
CAS 81720-08-3
Color White
Purity Grade Notes Research
Molecular Formula C27H42O20
Quantity 10 mg
Solubility Information H2O : 125 mg/mL (182.05 mM; Need ultrasonic)
SMILES O[C@H]1[C@@]([C@](C(CO)=C1)([H])[C@@H]2O[C@]([C@@H]([C@@H](O)[C@@H]3O)O)([H])O[C@@H]3CO)(C=CO2)O[C@@](O[C@H](CO)[C@@H](O)[C@@H]4O)([H])[C@@H]4O[C@]([C@@H]([C@@H](O)[C@@H]5O)O)([H])O[C@@H]5CO
Molecular Weight (g/mol) 686.61
Formula Weight 686.61
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.