missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Regulatory protein GAL4 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-76808JF646
This item is not returnable.
View return policy
Description
Regulatory protein GAL4 Polyclonal antibody specifically detects Regulatory protein GAL4 in Yeast samples. It is validated for Western Blot
Specifications
| Regulatory protein GAL4 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Partial recombinant protein made to a N terminal portion of the yeast GAL4 protein (amino acids 1-147) [UniProt P04386] | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 646 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Yeast | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction