missing translation for 'onlineSavingsMsg'
Learn More

Regulatory protein GAL4 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18411968 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411968 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411968 Supplier Novus Biologicals Supplier No. NBP276808AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Regulatory protein GAL4 Polyclonal antibody specifically detects Regulatory protein GAL4 in Yeast samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Regulatory protein GAL4
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias GAL4;GAL81;YPL248C
Host Species Rabbit
Immunogen Partial recombinant protein made to a N terminal portion of the yeast GAL4 protein (amino acids 1-147) [UniProt P04386]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Yeast
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.