missing translation for 'onlineSavingsMsg'
Learn More

Regucalcin Antibody (4B9), Novus Biologicals™

Product Code. 18355958 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18355958 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18355958 Supplier Novus Biologicals Supplier No. H00009104M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Regucalcin Monoclonal antibody specifically detects Regucalcin in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Regucalcin
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 4B9
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_004674
Gene Alias EC 3.1.1.17, Gluconolactonase, GNL, RCsenescence marker protein-30, regucalcin (senescence marker protein-30), Senescence marker protein 30, SMP-30, SMP30regucalcin
Host Species Mouse
Immunogen RGN (NP_004674.1, 200 a.a. ~ 299 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EQIPDGMCIDAEGKLWVACYNGGRVIRLDPVTGKRLQTVKLPVDKTTSCCFGGKNYSEMYVTCARDGMDPEGLLRQPEAGGIFKITGLGVKGIAPYSYAG
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9104
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.