missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Regorafenib Hydrochloride, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Regorafenib Hydrochloride (BAY 73-4506 hydrochloride) is a multi-target inhibitor for VEGFR1/2/3, PDGFRβ, Kit, RET and Raf-1 with IC50s of 13/4.2/46, 22, 7, 1.5 and 2.5 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | Regorafenib Hydrochloride |
| CAS | 835621-07-3 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H16Cl2F4N4O3 |
| Quantity | 10 mg |
| Synonym | BAY 73-4506 hydrochloride |
| Solubility Information | DMSO : ≥ 5.6 mg/mL (10.78 mM) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | ClC1=C(C(F)(F)F)C=C(NC(NC2=C(F)C=C(OC3=CC=NC(C(NC)=O)=C3)C=C2)=O)C=C1.Cl |
| Molecular Weight (g/mol) | 519.28 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction