missing translation for 'onlineSavingsMsg'
Learn More

Regadenoson, MedChemExpress

Product Code. 16493776
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493776 25 mg 25mg
16490738 5 mg 5mg
16468308 10 mg 10mg
16444197 50 mg 50mg
16410376 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16493776 Supplier Medchem Express Supplier No. HYA0168
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Regadenoson (CVT-3146) is a potent and selective A2A adenosine receptor agonist, with Kis of 290 and 1120 nM for rat and pig adenosine A2A receptor, respectively.

Regadenoson is selective for the adenosine A2A receptor over adenosine A1 and A2B receptors, and shows 13-fold selectivity over the human adenosine A1 receptor. Regadenoson is a vasodilator stress agent has shifted the landscape of vasodilator myocardial perfusion imaging. Regadenoson increases blood-brain barrier (BBB) permeability in rodents.

Specifications

Chemical Name or Material Regadenoson
CAS 313348-27-5
Color White
Purity Grade Notes Research
Molecular Formula C15H18N8O5
Quantity 25 mg
Synonym CVT-3146
Solubility Information DMSO : 50 mg/mL (128.09 mM; Need ultrasonic)
SMILES OC[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=C(N4N=CC(C(NC)=O)=C4)N=C3N)O1)O)O
Molecular Weight (g/mol) 390.35
Formula Weight 390.35
Percent Purity 98.35%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.