missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ REG4 Monoclonal Antibody (200214)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA560977

Product Code. 30533817

  • £490.00 / 500µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

Reg IV is part of the regenerating gene family within the C-type lectin superfamily. This family is involved in liver, pancreatic, gastric and intestinal cell proliferation and differentiation. Reg IV is a 158-amino acid secretory protein implicated in cell regeneration and/or survival with a definite growth stimulating effect on pancreatic beta cells. It is highly expressed in colorectal, gastric, prostate and other types of cancer. Reg IV-positive tumor cells display different phenotypes including mucus-secreting, enterocyte-like, and undifferentiated.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

REG4
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q9BYZ8
REG4
E. coli-derived recombinant human Reg4, Asp23-Pro158.
500 μg
Primary
Human
Antibody
IgG2b
Western Blot
200214
Unconjugated
REG4
2010002L15Rik; Gastrointestinal secretory protein; GISP; GISPREG-4; Reg IV; REG4; REG-4; regenerating family member 4; regenerating gene type IV; regenerating islet-derived family, member 4; regenerating islet-derived protein 4; Regenerating islet-derived protein IV; REG-IV; REG-like protein; RELP
Mouse
Protein A/G
RUO
83998
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.