missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Reg4 Polyclonal antibody specifically detects Reg4 in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | Reg4 |
| Applications | ELISA |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Human Reg4 (Accession#: NP_001152824.1; Met 1-Pro 158) |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?