missing translation for 'onlineSavingsMsg'
Learn More

Reg4 Antibody, FITC, Novus Biologicals™

Product Code. 18340105 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18340105 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18340105 Supplier Novus Biologicals Supplier No. NBP299734F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reg4 Polyclonal specifically detects Reg4 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Reg4
Applications ELISA
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias Gastrointestinal secretory protein, GISPREG-4, Reg IV, regenerating gene type IV, regenerating islet-derived family, member 4, regenerating islet-derived protein 4, Regenerating islet-derived protein IV, REG-IV, RELPREG-like protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Reg4 (NP_001152824.1; Met 1-Pro 158).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 83998
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.