missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reg4 Antibody (244), DyLight 550, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89832R
This item is not returnable.
View return policy
Description
Reg4 Monoclonal specifically detects Reg4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Reg4 | |
| Monoclonal | |
| DyLight 550 | |
| Gastrointestinal secretory protein, GISPREG-4, Reg IV, regenerating gene type IV, regenerating islet-derived family, member 4, regenerating islet-derived protein 4, Regenerating islet-derived protein IV, REG-IV, RELPREG-like protein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Reg4 (NP_001152824.1; Met 1-Pro 158). | |
| 0.1 mL | |
| Immunology | |
| 83998 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 244 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction