missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reg4 Antibody (244), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89832APC
394.40 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Reg4 Monoclonal antibody specifically detects Reg4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Reg4 | |
| Monoclonal | |
| APC | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| 244 | |
| Immunohistochemistry-Paraffin | |
| Gastrointestinal secretory protein, GISPREG-4, Reg IV, regenerating gene type IV, regenerating islet-derived family, member 4, regenerating islet-derived protein 4, Regenerating islet-derived protein IV, REG-IV, RELPREG-like protein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Reg4 (Accession#: NP_001152824.1; Met 1-Pro 158). | |
| 0.1 mL | |
| Immunology | |
| 83998 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction