missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ REG3G Polyclonal Antibody
GREENER_CHOICE

Product Code. 17712194
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17712194 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17712194 Supplier Invitrogen™ Supplier No. PA5120793

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: BxPC-3, U-87MG, Rat skeletal muscle. The target is usually found in the following locations: Cytoplasm, Secreted. Immunogen sequence: EVAKKDAPSS RSSCPKGSRA YGSYCYALFS VSKNWYDADM ACQKRPSGHL VSVLSGAEAS FLSSMIKSSG NSGQYVWIGL HDPTLGYEPN RGGWEWSNAD VMNYINWETN PSSSSGNHCG TLSRASGFLK WRENYCNLEL PYVCKFKA.

REG3G, also known as regenerating islet-derived 3 gamma, is a gene encoding an antimicrobial peptide crucial for maintaining intestinal homeostasis and regulating host-microbiome interactions. The protein produced by REG3G belongs to the C-type lectin family and is primarily expressed in the gut, where it plays a significant role in defending against bacterial infections. REG3G helps maintain gut barrier function and immune responses, acting in collaboration with interleukin 22 (IL-22), which regulates its expression. This gene is downregulated during chronic ethanol exposure, suggesting an influence on susceptibility to alcoholic liver disease. REG3G is also involved in glucose metabolism, with evidence showing that its administration improves glucose tolerance, potentially through actions mediated by its receptor, Extl3. Furthermore, REG3G has been identified as a nociceptor-enriched hormone, which protects against endotoxic shock by modulating both local and systemic immunity. This diverse functionality underscores the gene's importance in immune modulation, gut physiology, and metabolic processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REG3G
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene REG3G
Gene Accession No. O09049, P42854, Q6UW15
Gene Alias AI449515; generating islet-derived, mouse homolog 3 gamma; LPPM429; Pancreatitis-associated protein 1B; pancreatitis-associated protein 3; Pancreatitis-associated protein IB; PAP IB; PAP1B; PAP-1B; Pap3; PAPIB; PAPIII; REG III; Reg III-gamma; REG3G; REG3gamma; REG-3gamma; REG-3-gamma; regenerating family member 3 gamma; regenerating gene III; regenerating islet-derived 3 gamma; regenerating islet-derived protein 3 gamma; regenerating islet-derived protein 3-gamma; Regenerating islet-derived protein 3-gamma 15 kDa form; Regenerating islet-derived protein 3-gamma 16.5 kDa form; Regenerating islet-derived protein III-gamma; REG-III; RegIII (gamma); UNQ429; UNQ429/PRO162
Gene Symbols REG3G
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 27-174 of mouse REG3G (NP_0353901).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130120, 19695, 24620
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.