missing translation for 'onlineSavingsMsg'
Learn More

Reg3G Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18340742 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18340742 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18340742 Supplier Novus Biologicals Supplier No. NBP306396AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reg3G Polyclonal antibody specifically detects Reg3G in Human samples. It is validated for ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Reg3G
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias LPPM429, MGC118999, MGC119001, Pancreatitis-associated protein 1B, Pancreatitis-associated protein IB, PAP IB, PAP-1B, PAP1BMGC118998, PAPIB, Reg III-gamma, REG-3-gamma, regenerating gene III, regenerating islet-derived 3 gamma, regenerating islet-derived protein 3-gamma, Regenerating islet-derived protein III-gamma, REG-III, UNQ429
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Reg3G (NP_001008388.1; Met 1-Asp 175).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130120
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.