missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ REG3A Polyclonal Antibody
GREENER_CHOICE

Product Code. 17179423
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17179423 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17179423 Supplier Invitrogen™ Supplier No. PA5111509

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: YVWIGLHDPT QGTEPNGEGW EWSSSDVMNY FAWERNPSTI SSPGHCASLS RSTAFL Highest antigen sequence indentity to the following orthologs: Mouse - 70%, Rat - 70%.

REG3A encodes a pancreatic secretory protein that may be involved in cell proliferation or differentiation. It has similarity to the C-type lectin superfamily. The enhanced expression of this protein is observed during pancreatic inflammation and liver carcinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REG3A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Reg3a
Gene Accession No. Q06141
Gene Alias AV063448; generating islet-derived, mouse homolog 3 alpha; hepatocarcinoma-intestine-pancreas; hepatointestinal pancreatic protein; HIP; HIP/PAP; human proislet peptide; INGAP; islet of Langerhans regenerating protein 3; Lithostathine 3; pancreatic beta cell growth factor; pancreatitis-associated protein 1; Pancreatitis-associated protein 2; PAP; PAP homologous protein; PAP1; Pap2; PAP-H; PapII; PBCGF; proliferation-inducing protein 34; proliferation-inducing protein 42; REG 3; REG 3a; REG 3alpha; Reg IIIalpha; Reg III-alpha; REG3; REG3 alpha; Reg3a; REG3alpha; REG-3alpha; REG-3-alpha; regeneratin; regenerating family member 3 alpha; regenerating islet-derived 3 alpha; regenerating islet-derived protein 3 alpha; regenerating islet-derived protein 3-alpha; Regenerating islet-derived protein 3-alpha 15 kDa form; Regenerating islet-derived protein 3-alpha 16.5 kDa form; Regenerating islet-derived protein III-alpha; regIII; REG-III; RegIII (alpha)
Gene Symbols Reg3a
Host Species Rabbit
Immunogen Recombinant protein fragment of Human REG3A (Product #RP-105618)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5068
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.