missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reg3A Antibody (019), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89856H
This item is not returnable.
View return policy
Description
Reg3A Monoclonal specifically detects Reg3A in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Reg3A | |
| Monoclonal | |
| HRP | |
| hepatocarcinoma-intestine-pancreas, HIPpancreatitis-associated protein, Human proislet peptide, pancreatic beta cell growth factor, Pancreatitis-associated protein 1, PAP homologous protein, PAP1FLJ18565, PAPPAP-H, PBCGF, proliferation-inducing protein 34, proliferation-inducing protein 42, Reg III-alpha, REG3, REG-3-alpha, regenerating islet-derived 3 alpha, regenerating islet-derived protein 3-alpha, Regenerating islet-derived protein III-alpha, REG-III | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Reg3A (Q06141-1; Met 1-Asp 175). | |
| 0.1 mL | |
| Primary | |
| No cross-reactivity in ELISA with: Human REG3G Mouse REG3A Human REG1B | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 19 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 5068 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction