missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ REG1B Recombinant Rabbit Monoclonal Antibody (103), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552301
This item is not returnable.
View return policy
Description
REG1B might act as an inhibitor of spontaneous calcium carbonate precipitation. REG1B may be associated with neuronal sprouting in brain, and with brain and pancreas regeneration.This gene is a type I subclass member of the Reg gene family. The Reg gene family is a multigene family grouped into four subclasses, types I, II, III and IV based on the primary structures of the encoded proteins. This gene encodes a protein secreted by the exocrine pancreas that is highly similar to the REG1A protein. The related REG1A protein is associated with islet cell regeneration and diabetogenesis, and may be involved in pancreatic lithogenesis. Reg family members REG1A, REGL, PAP and this gene are tandemly clustered on chromosome 2p12 and may have arisen from the same ancestral gene by gene duplication. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
Specifications
| REG1B | |
| Recombinant Monoclonal | |
| 100 μg/mL | |
| PBS with no preservative | |
| P48304 | |
| REG1B | |
| Recombinant Human REG1B protein (Gln 23-Asn166). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 103 | |
| Unconjugated | |
| REG1B | |
| lithostathine 1 beta; lithostathine-1-beta; Pancreatic stone protein 2; PSP-2; PSPS2; REG1B; REG-1-beta; regenerating family member 1 beta; regenerating islet-derived 1 beta; regenerating islet-derived 1 beta (pancreatic stone protein, pancreatic thread protein); regenerating islet-derived protein 1-beta; Regenerating protein I beta; REGH; REGI-BETA; REGL; secretory pancreatic stone protein 2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 5968 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction