missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ REG1B Recombinant Rabbit Monoclonal Antibody (103), HRP, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£142.00 - £444.00
Specifications
| Antigen | REG1B |
|---|---|
| Clone | 103 |
| Concentration | 100 μg/mL |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019412
![]() |
Invitrogen™
MA552303 |
100 μg |
£444.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019218
|
Invitrogen™
MA552304 |
20 μg |
£142.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
REG1B Recombinant Monoclonal antibody specifically detects REG1B in Human samples. It is validated for ELISASpecifications
| REG1B | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| lithostathine 1 beta; lithostathine-1-beta; Pancreatic stone protein 2; PSP-2; PSPS2; REG1B; REG-1-beta; regenerating family member 1 beta; regenerating islet-derived 1 beta; regenerating islet-derived 1 beta (pancreatic stone protein, pancreatic thread protein); regenerating islet-derived protein 1-beta; Regenerating protein I beta; REGH; REGI-BETA; REGL; secretory pancreatic stone protein 2 | |
| Recombinant Human REG1B protein (Gln 23-Asn166). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| REG1B |
| 103 | |
| ELISA | |
| HRP | |
| Rabbit | |
| Human | |
| P48304 | |
| 5968 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title