missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reg1B Antibody (029), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89939CL1
This item is not returnable.
View return policy
Description
Reg1B Monoclonal antibody specifically detects Reg1B in Human samples. It is validated for ELISA
Specifications
| Reg1B | |
| Monoclonal | |
| CoraFluor 1 | |
| lithostathine-1-beta, Pancreatic stone protein 2, pancreatic threadprotein), PSPS2PSP-2, regenerating islet-derived 1 beta, Regenerating islet-derived protein 1-beta, Regenerating protein I beta, REGH, REGLREG-1-beta, secretory pancreatic stone protein 2 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Reg1B (Accession#: NP_006498.1; Met 1-Asn 166). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 029 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 5968 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction