missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Refractive Index Certified Reference Material at 20 °C, 25 °C and 30 °C (Nominal RI Value 1.4573 at 25 °C), UKAS ISO 17025 / ISO 17034 Certified, in Tamper-Evident Glass Vials, 10 mL, Paragon Scientific™
Description
- Refractive Index Certified Reference Materials at 20 °C, 25 °C and 30 °C
- Certified in strict accordance to ISO 17025 and ISO 17034 under our UKAS accreditation
- Highest level of accreditation guarantee, providing the most credible certified data available worldwide
- All measurements are fully traceable to NIST and international protocols
- Low levels of uncertainty, ensuring maximum accuracy of data at hand and dependable results
- Available in single 10 mL volume tamper-evident glass bottles or as a multi-pack of 5 per material (includes set of disposable pipettes)
- 12 month shelf life
- Manufactured in the United Kingdom
Specifications
Specifications
| Quantity | 10 mL |
Safety and Handling
Product Identifier
- Refractive Index Standard at 20 °C
- Danger
- Aspiration toxicity Category 1
- H304-May be fatal if swallowed and enters airways.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : petroleum distilates
Nominal values quoted. Certified Values will vary from batch to batch.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?